HPLC Analysis of Benzoic Acid Deriviatives on Ascentis® RP-Amide
HPLC Analysis of Benzoic Acid Deriviatives on Ascentis® RP-Amide
Conditions
column Ascentis RP-Amide, 15 cm x 4.6 mm I.D., 5 µm particles (565324-U)
column temp. 30 C
mobile phase [A] 0.1% TFA in water; [B] 0.1% TFA in acetonitrile; (70:30, A:B)
flow rate 1.0 ml/min
sample as indicated in 0.1% TFA in water
injection 10 µl
detector UV, 220 nm
Description
Categories Analytical Chromatography, Organic Acids
Featured Industry Chemicals and Industrial Polymers