HPLC Analysis of Benzoin Enantiomers on Astec® (R,R) P-CAP™-DP
HPLC Analysis of Benzoin Enantiomers on Astec® (R,R) P-CAP™-DP
-
Conditions
column Astec (R,R) P-CAP-DP, 25 cm x 4.6 mm I.D., 5 µm particles (35024AST)
column temp. 25 °C
mobile phase [A] hexane; [B] ethanol; (80:20, A:B)
flow rate 1 mL/min
sample 1 mg/mL in methanol
injection 2 µL
detector UV, 230 nm
Description
Categories Analytical Chromatography, Chiral Applications, Antibiotics, Antimicrobials, Pharmaceutical (small molecule), Antibiotics/Antimicrobials, All Pharmaceutical Applications
Featured Industry Pharmaceutical (small molecule)
Legal Information P-CAP is a trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC