HPLC Analysis of Benzoin Methyl Ether Enantiomers on Astec® CHIROBIOTIC® V
HPLC Analysis of Benzoin Methyl Ether Enantiomers on Astec® CHIROBIOTIC® V
Conditions
column CHIROBIOTIC V, 25 cm x 4.6 mm I.D., 5 µm particles (11024AST)
column temp. 25 °C
mobile phase [A] 1% triethylamine, pH 7.0; [B] THF; (90:10, A:B)
flow rate 1 mL/min
sample 1 mg/mL in methanol
injection 2 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Chiral Applications, Organic Chemicals, Chemicals and Industrial Polymers, Alcohols, Phenols & Diols, Other Organic Compound Applications
Featured Industry Vitamins, Nutraceuticals, and Natural Products
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC