HPLC Analysis of Beta Blockers on Ascentis® Si (Silica)
HPLC Analysis of Beta Blockers on Ascentis® Si (Silica)
Conditions
column Ascentis Si, 15 cm x 4.6 mm I.D., 5 µm particles (581512-U)
column temp. 35 °C
mobile phase [A] 0.1% ammonium acetate in water (pH unadjusted); [B] 0.1% ammonium acetate in acetonitrile; (15:85, A:B)
flow rate 1 mL/min
sample as indicated in 0.1% ammonium acetate in 10:90:water:acetonitrile
injection 10 µL
detector UV, 225 nm
Description
Categories Analytical Chromatography, Clinical, antihypertensives, beta-blockers, Antihypertensives
Featured Industry Clinical
Pharmaceutical (small molecule)