HPLC Analysis of Beta-Receptor Agonist Enantiomers on Astec® CHIROBIOTIC® T
HPLC Analysis of Beta-Receptor Agonist Enantiomers on Astec® CHIROBIOTIC® T
Conditions
column CHIROBIOTIC T, 25 cm x 4.6 mm, 5 µm particles (12024AST)
column temp. 25 °C
mobile phase 15 mM ammonium formate in methanol
flow rate 1 mL/min
detector UV, 220 nm
Description
Categories Analytical Chromatography, Clinical, Forensics and Toxicology, Chiral Applications, antihypertensives, Clinical, Forensic & Toxicology, bronchodilators, Bronchodilators, Pharmaceutical (small molecule), All Pharmaceutical Applications, Therapeutic Drug Monitoring
Featured Industry Clinical
Forensics and Toxicology
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC