Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Beverage Additives in Diet Cola on Ascentis® Express RP-Amide
HPLC Analysis of Beverage Additives in Diet Cola on Ascentis® Express RP-Amide
Conditions
column Ascentis Express RP-Amide, 10 cm x 3.0 mm I.D., 2.7 µm particles (53918-U)
column temp. 40 C
mobile phase [A] 20 mM ammonium acetate, pH 4.7 with acetic acid; [B] acetonitrile; (92:8, A:B)
flow rate 0.6 ml/min
sample 500 - 1000 µg/mL in 95:5, buffer: acetonitrile
injection 1 µl
detector UV, 214 nm
Description
Analysis Note This application demonstrates the suitability of Ascentis Express RP-Amide for the efficient separation of common drink additives used in diet cola.
Categories Analytical Chromatography, Food & Beverage Analysis, Carboxylic acids, Beverage Testing, Stiµlants, Sweeteners, Soft Drinks
Featured Industry Food and Beverages