Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Bilirubin and Biliverdin on Discovery® C18
HPLC Analysis of Bilirubin and Biliverdin on Discovery® C18
Conditions
column Discovery C18, 10 cm x 2.1 mm I.D., 5 µm particles (569220-U)
column temp. ambient
mobile phase [A] 10 mM ammonium acetate (pH unadjusted) [B] 90:10, methanol:(100 mM ammonium acetate)
flow rate 0.2 ml/min
sample as indicated (in 50% MeOH, 1% NH4OH)
injection 1 µl
detector UV, 405 nm
Description
Categories Analytical Chromatography, Clinical, Pharmaceutical Analysis, Antioxidants, antioxidants, Proteins
Featured Industry Clinical
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC