HPLC Analysis of Binaphthol Enantiomers on Astec® CYCLOBOND® I 2000 DNP
HPLC Analysis of Binaphthol Enantiomers on Astec® CYCLOBOND® I 2000 DNP
Conditions
column CYCLOBOND I 2000 DNP, 25 cm x 4.6 mm I.D., 5 µm particles (25024AST)
column temp. 23 °C
mobile phase [A] 0.1% triethyl ammonium acetate, pH 4.1; [B] acetonitrile; (75:25, A:B)
flow rate 1.0 mL/min
injection 5 µL
detector UV, 230 nm
Description
Categories Analytical Chromatography, Phenols
Featured Industry Chemicals and Industrial Polymers
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CYCLOBOND is a registered trademark of Sigma-Aldrich Co. LLC