HPLC Analysis of Bovine Serum Albumin (BSA) on Zenix® SEC-300 versus SRT® SEC-300, Effect of Particle Size on Resolution
HPLC Analysis of Bovine Serum Albumin (BSA) on Zenix® SEC-300 versus SRT® SEC-300, Effect of Particle Size on Resolution
Conditions
column SRT SEC-300, 30 cm x 7.8 mm I.D., 5 µm, 300 Angstrom particles (Z777051)
column Zenix SEC-300, 30 cm x 7.8 mm I.D., 3 µm, 300 Angstrom particles (Z777033)
column temp. ambient
mobile phase 150 mM sodium phosphate, pH 7
flow rate 1 ml/min
sample 5 mg/mL, BSA
injection 10 µl
detector UV, 214 nm
Description
Categories Analytical Chromatography, Clinical, Proteins, proteins
Featured Industry Life Science and Biopharma
Pharmaceutical (small molecule)
Legal Information SRT is a registered trademark of Sepax Technologies, Zenix is a registered trademark of Sepax Technologies