HPLC Analysis of Bromacil Enantiomers on Astec® CHIROBIOTIC® T
HPLC Analysis of Bromacil Enantiomers on Astec® CHIROBIOTIC® T
Conditions
column CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 µm particles (12024AST)
column temp. 23 C
mobile phase [A] water; [B] ethanol; (60:40, A:B)
flow rate 0.6 ml/min
sample 1 mg/mL in methanol
injection 2 µl
detector UV, 215 nm
Description
Categories Analytical Chromatography, Chiral Applications, Pesticides & Herbicides, Environmental, All Environmental Applications, Herbicides
Featured Industry Pharmaceutical (small molecule)
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC