HPLC Analysis of Bufalin Toxin on Discovery® BIO Wide Pore C18
HPLC Analysis of Bufalin Toxin on Discovery® BIO Wide Pore C18
Conditions
column Discovery BIO Wide Pore C18, 25 cm x 4.6 mm I.D., 5 µm particles (568223-U)
column temp. ambient
mobile phase [A] 0.1% TFA in water; [B] 0.1% TFA in acetonitrile:water (3:1, A:B)
flow rate 1.3 ml/min
injection 10 muLbufalin, 0.5 mg/mL in acetonitrile
detector UV, 298 nm
Description
Categories Analytical Chromatography, Forensics and Toxicology, steroids, Steroids
Featured Industry Forensics and Toxicology
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC