HPLC Analysis of Bupivacaine Enantiomers on Astec® CHIROBIOTIC® V2, Formate Mobile Phase
HPLC Analysis of Bupivacaine Enantiomers on Astec® CHIROBIOTIC® V2, Formate Mobile Phase
Conditions
column CHIROBIOTIC V2, 25 cm x 4.6 mm I.D., 5 µm particles (15024AST)
column temp. 20 C
mobile phase [A] 20 mM ammonium formate, pH 4.1; [B] methanol; (5:95, A:B)
flow rate 1 ml/min
sample 1 mg/mL in methanol
injection 5 µl
detector UV, 220 nm
Description
Categories Analytical Chromatography, Chiral Applications, Anesthetics, Pharmaceutical (small molecule), All Pharmaceutical Applications, Anaesthetics
Featured Industry Pharmaceutical (small molecule)
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC