HPLC Analysis of Caffeine and Metabolites on Ascentis® RP-Amide
HPLC Analysis of Caffeine and Metabolites on Ascentis® RP-Amide
Conditions
column Ascentis RP-Amide, 15 cm x 4.6 mm I.D., 5 µm particles (565324-U)
column temp. 30 °C
mobile phase [A] 0.1% ammonium acetate in water, pH 4.5 with acetic acid; [B] acetonitrile
gradient 2 to 8% B in 3 min; to 20% B in 9 min; held for 2 min; to 2% B in 0.1 min; held for 0.9 min
flow rate 1.0 mL/min
sample as indicated in 0.1% ammonium acetate in water (pH 4.5 with acetic acid)
injection 10 µL
detector UV, 275 nm
Description
Categories Analytical Chromatography, Clinical, stiµLants, StiµLants, drugs of abuse, Drugs of Abuse
Featured Industry Clinical