HPLC Analysis of Caffeine Metabolites and Analogs on Discovery® C18
HPLC Analysis of Caffeine Metabolites and Analogs on Discovery® C18
Conditions
column Discovery C18, 15 cm x 4.6 mm I.D., 5 µm particles (504955)
column temp. 35 °C
mobile phase [A] 1% acetic acid in water; [B] methanol; (92:8, A:B)
flow rate 1.0 mL/min
detector UV, 254 nm
Description
Categories Analytical Chromatography, Clinical, drugs of abuse, stimulants, Stimulants, Drugs of Abuse
Featured Industry Clinical
Vitamins, Nutraceuticals, and Natural Products
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC