HPLC Analysis of Cannabinoids on Ascentis® C18
HPLC Analysis of Cannabinoids on Ascentis® C18
Conditions
column Ascentis C18, 10 cm x 4.6 mm I.D., 3 µm particles (581321-U)
column temp. 35 °C
mobile phase [A] 10 mM ammonium acetate (pH 4.5 with acetic acid); [B] acetonitrile; (25:75, A:B)
flow rate 1 mL/min
sample 30 µg/mL each in mobile phase
injection 10 µL
detector UV, 220 nm
Description
Categories Analytical Chromatography, Forensics and Toxicology, drugs of abuse, analgesics, Analgesics
Featured Industry Forensics and Toxicology