HPLC Analysis of Capsacinoids in a Capsicum annuum L. extract.on Ascentis® Express C18
HPLC Analysis of Capsacinoids in a Capsicum annuum L. extract.on Ascentis® Express C18
Conditions
column Ascentis Express C18, 15 cm x 4.6 mm I.D., 2.7 µm (53829-U)
mobile phase [A] 0.075% acetic acid, pH 3; [B] 0.075% acetic acid in acetonitrile; (40:60; A:B)
flow rate 1 mL/min
injection 5 µL
detector UV, 280 nm
Description
Analysis Note Two grams of Capsicum annuum L. pericarp, placenta, and seeds were separately homogenized and extracted with 50 mL of water/methanol (25/75, v/v) for 20 minutes at 100 °C. The extracts were filtered and evaporated at 45 °C then resuspended in 1 mL of water/methanol (1/1, v/v).
Categories Analytical Chromatography, Capsaicinoids
Featured Industry Food and Beverages
Legal Information Ascentis is a registered trademark of Sigma-Aldrich Co. LLC
suitability suitable for HPLC