HPLC Analysis of Carbamate and Urea Pesticides on SUPELCOSIL™ LC-8
HPLC Analysis of Carbamate and Urea Pesticides on SUPELCOSIL™ LC-8
Conditions
column SUPELCOSIL LC-8, 15 cm x 4.6 mm I.D., 5 µm particles (58220-U)
column temp. 35 °C
mobile phase [A] acetonitrile; [B] water
gradient 18:82 to 65:35 in 9 min, hold 3 min
flow rate 2 mL/min
injection 10 µL
detector UV, 240 nm
Description
Categories Analytical Chromatography, Environmental Analysis, Pesticides & Herbicides, Organics, Chromatograms, HPLC Chromatograms
Featured Industry Environmental
Legal Information SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC