HPLC Analysis of Carbamazepine on Ascentis® Express ES-Cyano
HPLC Analysis of Carbamazepine on Ascentis® Express ES-Cyano
Conditions
column Ascentis Express ES-Cyano, 10 cm x 4.6 mm I.D., 2.7 µm particles (53491-U)
column temp. 30 °C
mobile phase water:methanol:tetrahydrofuran (85:12:3); formic acid, triethylamine
flow rate 1.25 mL/min
injection 20 µL
detector UV, 230 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Anticonvulsant/antiepiletic/antiseizure