HPLC Analysis of Cardiac Drugs on Ascentis® C8
HPLC Analysis of Cardiac Drugs on Ascentis® C8
Conditions
column Ascentis C8, 15 cm x 2.1 mm I.D., 3 µm particles (581402-U)
column temp. 30 °C
mobile phase 0.1% ammonium acetate:acetonitrile with 0.1% ammonium acetate, (47:53)
flow rate 0.2 mL/min
sample as indicated in mobile phase
injection 2 µL
detector UV, 220 nm
Description
Categories Analytical Chromatography, Antihypertensives