HPLC Analysis of Carnitine Enantiomers on Astec® CHIROBIOTIC® TAG
HPLC Analysis of Carnitine Enantiomers on Astec® CHIROBIOTIC® TAG
Conditions
column CHIROBIOTIC TAG, 25 cm x 4.6 mm I.D., 5 µm particles (14024AST)
column temp. 35 °C
mobile phase [A] 5 mM ammonium acetate, pH 6; [B] methanol; (25:75, A:B)
flow rate 0.6 mL/min
sample 1 µg/mL in mobile phase
injection 5 µL
detector ESI(+)-MS (m/z = 162)
Description
Categories Analytical Chromatography, Chiral Applications, Amino Acids, Life Science & Biopharma, All Biochemical Applications, All Amino Acids, Native Amino Acids
Featured Industry Life Science and Biopharma
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC