HPLC Analysis of Carotene Compounds on Ascentis® RP-Amide
HPLC Analysis of Carotene Compounds on Ascentis® RP-Amide
Conditions
column Ascentis RP-Amide, 15 cm x 4.6 mm I.D., 5 µm particles (565324-U)
column temp. 35 °C
mobile phase 100% acetonitrile
flow rate 1.0 mL/min
sample 10 µg/mL each in 100% acetonitrile
injection 5 µL
detector MS, ESI(+) in selective ion monitoring (SIR) mode
Description
Categories Analytical Chromatography, Vitamins and Natural products
Featured Industry Food and Beverages
Vitamins, Nutraceuticals, and Natural Products