HPLC Analysis of Carotenoids on Ascentis® Si (Silica), Normal Phase
HPLC Analysis of Carotenoids on Ascentis® Si (Silica), Normal Phase
Conditions
column Ascentis Si, 15 cm x 4.6 mm I.D., 5 µm particles (581512-U)
column temp. 30 °C
mobile phase [A] hexane: [B] ethyl acetate: [C] isopropanol; (85:10:5, A:B:C)
flow rate 1.0 mL/min
sample as indicated in hexane
injection 10 µL
detector UV-VIS, 450 nm
Description
Categories Analytical Chromatography, Vitamins and Natural products
Featured Industry Vitamins, Nutraceuticals, and Natural Products