Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Cephalsporin Antibiotics on Ascentis® RP-Amide
HPLC Analysis of Cephalsporin Antibiotics on Ascentis® RP-Amide
Conditions
column Ascentis RP-Amide, 15 cm x 4.6 mm I.D., 5 µm particles (565324-U)
column temp. 35 C
mobile phase [A] 10 mM monobasic potassium phosphate (pH 3.0 with phosphoric acid): [B] acetonitrile; (93:7, A:B)
flow rate 1 ml/min
sample 50 µg/mL each in mobile phase
injection 10 µl
detector UV, 254 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Antibiotics, Antimicrobials, Antibiotics