HPLC Analysis of Cetirizine and Hydroxyzine on Ascentis® Express F5
HPLC Analysis of Cetirizine and Hydroxyzine on Ascentis® Express F5
Conditions
column Ascentis Express F5, 10 cm x 3.0 mm I.D., 2.7 µm particles (53578-U)
column temp. 25 C
mobile phase 5 mM ammonium formate pH 3.0 with formic acid in 90:10, acetonitrile:water
flow rate 0.6 ml/min
pressure 1300 psi (90 bar)
sample 50 µg/mL in 90:10, acetonitrile:water
injection 2 µl
detector UV, 254 nm
Description
Categories Analytical Chromatography, Antihistamines