Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Chlorhexidine and Robenidine on Ascentis® Phenyl
HPLC Analysis of Chlorhexidine and Robenidine on Ascentis® Phenyl
Conditions
column Ascentis Phenyl 15 cm x 4.6 mm I.D., 5 µm particles (581616-U)
column temp. 35 C
mobile phase [A] water; [B] acetonitrile; [C] 500 mM ammonium formate, pH 3.0 with formic acid
gradient 10% C constant; 65% A, 25% B to 0% A, 90% B in 7 min; held for 2 min; to 65% A, 25% B in 0.5 min
flow rate 1.0 ml/min
sample 100 ug/mL each in 70:30, 25 mM ammonium formate pH 3.0:acetonitrile
injection 10 µl
detector UV, 273 nm
Description
Categories Analytical Chromatography, Antibiotics, Antimicrobials
Featured Industry Cosmetics, Personal Care, and Cleaning Products
Pharmaceutical (small molecule)