HPLC Analysis of Chlorthalidone Enantiomers on Astec® CHIROBIOTIC® T
HPLC Analysis of Chlorthalidone Enantiomers on Astec® CHIROBIOTIC® T
Conditions
column CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 µm particles (12024AST)
column temp. 25 °C
mobile phase [A] 10 mM ammonium acetate, pH 4.1; [B] methanol; (80:20, A:B)
flow rate 0.9 mL/min
sample 2 mg/mL in methanol
injection 5 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Chiral Applications, Antihypertensives, Pharmaceutical (small molecule), All Pharmaceutical Applications
Featured Industry Pharmaceutical (small molecule)
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC