HPLC Analysis of Chlorthalidone Enantiomers on Astec® CYCLOBOND® I 2000 HP-RSP
HPLC Analysis of Chlorthalidone Enantiomers on Astec® CYCLOBOND® I 2000 HP-RSP
Conditions
column CYCLOBOND I 2000 HP-RSP, 25 cm x 4.6 mm I.D., 5 µm particles (24024AST)
column temp. 25 °C
mobile phase [A] 10 mM ammonium acetate, pH 4.0; [B] acetonitrile; (85:15, A:B)
flow rate 1 mL/min
sample 5 mg/mL in methanol
injection 2 µL
detector UV, 235 nm
Description
Categories Analytical Chromatography, Chiral Applications, Antihypertensives, Pharmaceutical (small molecule), All Pharmaceutical Applications
Featured Industry Pharmaceutical (small molecule)
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CYCLOBOND is a registered trademark of Sigma-Aldrich Co. LLC