HPLC Analysis of Chlorzoxazone on Discovery® HS-PEG
HPLC Analysis of Chlorzoxazone on Discovery® HS-PEG
Conditions
column Discovery HS PEG, 15 cm x 4.6 mm I.D., 5 µm particles (567416-U)
column temp. 30 °C
mobile phase [A] 20 mM acetic acid in water (pH 4.5 with ammonium hydroxide):[B] acetonitrile; (70:30, A:B)
flow rate 1 mL/min
injection 10 µL
detector UV, 285 nm
Description
Categories Analytical Chromatography, Neurotransmitters, Drugs of Abuse
Featured Industry Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC