HPLC Analysis of Cholesterol-Lowering Drugs (Statins) on Discovery® Cyano
HPLC Analysis of Cholesterol-Lowering Drugs (Statins) on Discovery® Cyano
Conditions
column Discovery Cyano, 15 cm x 4.6mm I.D., 5 µm particles (59356-U)
column temp. 35 °C
mobile phase 60:40 water (10 mM ammonium acetate, pH 4.0 with acetic acid):acetonitrile
flow rate 1.0 mL/min
sample 50 µg/mL each in 70:30 water:acetonitrile
injection 5 µL
detector UV, 230 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Anticholinergics, Anticholesterolemics
Featured Industry Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC