HPLC Analysis of Ciprofibrate Enantiomers on Astec® CYCLOBOND® I 2000
HPLC Analysis of Ciprofibrate Enantiomers on Astec® CYCLOBOND® I 2000
Conditions
column CYCLOBOND I 2000, 25 cm x 4.6 mm I.D., 5 µm particles (20024AST)
column temp. ambient
mobile phase [A] acetonitrile; [B] methanol; [C] acetic acid; [D] triethylamine; (90:10:0.3:0.2, A:B:C:D)
flow rate 1 ml/min
sample 5 mg/mL in methanol
injection 1 µl
detector UV, 230 nm
Description
Categories Analytical Chromatography, Chiral Applications, Pharmaceutical (small molecule), Anticholesterolemics, All Pharmaceutical Applications, Anticholesterol Drugs
Featured Industry Pharmaceutical (small molecule)
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CYCLOBOND is a registered trademark of Sigma-Aldrich Co. LLC