HPLC Analysis of Cis-(+/-)-3,3a,8,8a-Tetrahydro-2H-indeno[1,2-d]oxazol-2-one Enantiomers on Astec® (R,R) P-CAP™-DP
HPLC Analysis of Cis-(+/-)-3,3a,8,8a-Tetrahydro-2H-indeno[1,2-d]oxazol-2-one Enantiomers on Astec® (R,R) P-CAP™-DP
Conditions
column Astec (R,R) P-CAP-DP, 25 cm x 4.6 mm I.D., 5 µm particles (35024AST)
column temp. 25 °C
mobile phase [A] acetonitrile; [B] methanol; (98:2, A:B)
flow rate 1 mL/min
sample 1 mg/mL in methanol
injection 1 µL
detector UV, 230 nm
Description
Categories Analytical Chromatography, Chiral Applications, Chemicals and Industrial Polymers, Oxygen compounds, Organic Chemicals, Other Organic Compound Applications
Featured Industry Chemicals and Industrial Polymers
Legal Information P-CAP is a trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC