HPLC Analysis of Citalopram Enantiomers on Astec® CHIROBIOTIC® V
HPLC Analysis of Citalopram Enantiomers on Astec® CHIROBIOTIC® V
Conditions
column CHIROBIOTIC V, 25 cm x 4.6 mm I.D., 5 µm particles (11024AST)
column temp. 20 °C
mobile phase [A] methanol; [B] acetic acid; [C] triethylamine; (99.8:0.1:0.1, A:B:C)
flow rate 0.5 mL/min
sample citalopram, 1 mg/mL in mobile phase
injection 5 µL
detector UV, 213 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Chiral Applications, Antidepressants, Pharmaceutical (small molecule), All Pharmaceutical Applications
Featured Industry Pharmaceutical (small molecule)
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC