HPLC Analysis of Clopidogrel Enatiomers on Astec® CYCLOBOND® I 2000 DNP
HPLC Analysis of Clopidogrel Enatiomers on Astec® CYCLOBOND® I 2000 DNP
Conditions
column CYCLOBOND® I 2000 DNP, 25 cm x 4.6 mm I.D., 5 µm particles (25024AST)
column temp. 35 °C
mobile phase [A] 20 mM ammonium acetate, pH 4.0; [B] acetonitrile; (80:20, A:B)
flow rate 1 mL/min
pressure 1100 psi (76 bar)
sample 0.2 mg/mL in methanol
injection 2 µL
detector UV, 220 nm
Description
Categories Analytical Chromatography, Cardiac Drugs, Anticoagulants
Featured Industry Pharmaceutical (small molecule)
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CYCLOBOND is a registered trademark of Sigma-Aldrich Co. LLC