Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Cold Remedy Components on Ascentis® Express 2.0 μm RP-Amide
HPLC Analysis of Cold Remedy Components on Ascentis® Express 2.0 μm RP-Amide
Conditions
column Ascentis Express RP-Amide, 10 cm x 2.1 mm I.D., 2.0 µm particles (51576-U)
column temp. 35 C
mobile phase [A] 25 mM ammonium phophate pH 2.7 with 30% Phosporic Acid; [B] acetonitrile; (93:7, A:B)
flow rate 0.2 ml/min
pressure 3162 psi (218 bar)
sample Acetaminophen, Codeine, Doxylamine, Pseudoephedrine, 50   µg/mL in 95:5 mobile phase A:mobile phase B
injection 1 µl
detector UV, 215 nm
Description
Analysis Note This application demonstrates the separation of cold remedy components on Ascentis Express 2.0 μm RP-Amide.  The polarity and H-bond donating groups of the stationary phase are well suited for this mixture of phenols and basic components.  This is an update of a previous application on Ascentis RP-Amide to reflect scalability of the RP-Amide chemistry to the core shell particle.
Categories Analytical Chromatography, Antihistamines, Decongestants