Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Cold Remedy Ingredients on Ascentis® RP-Amide
HPLC Analysis of Cold Remedy Ingredients on Ascentis® RP-Amide
Conditions
column Ascentis RP-Amide, 15 cm x 4.6 mm I.D., 5 µm particles (565324-U)
column temp. 30 C
mobile phase [A] 5 mM ammonium formate, pH 2.75 with formic acid; [B] acetonitrile
gradient 10 to 30% B in 15 min; held for 2 min; to 10% B in 0.1 min
flow rate 1.0 ml/min
sample as indicated in 5 mM ammonium formate (pH 2.75 with formic acid)
injection 10 µl
detector UV, 270 nm
Description
Categories Analytical Chromatography, Antihistamines