Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Column Test Mix on Discovery® BIO Wide Pore C18, Effect of HPLC Column I.D. on Sensitivity
HPLC Analysis of Column Test Mix on Discovery® BIO Wide Pore C18, Effect of HPLC Column I.D. on Sensitivity
Conditions
column Discovery BIO Wide Pore C18, 10 cm x various I.D., 3 µm particles
column temp. ambient
mobile phase [A] water; [B] acetonitrile; (65:35, A:B)
flow rate as indicated
sample acetophenone (0.1 mug), benzene (1 mug), toluene (1 mug)
injection as indicated
detector UV, 254 nm
Description
Categories Analytical Chromatography, Aromatics
Featured Industry Life Science and Biopharma
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC