Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Cortocisteroids in Urine on Discovery® HS F5 after SPE using Discovery® DSC-CN
HPLC Analysis of Cortocisteroids in Urine on Discovery® HS F5 after SPE using Discovery® DSC-CN
Conditions
sample preparation SPE (Solid Phase Extraction)
sample/matrix human urine spiked with corticosteroids at 0.5 and 1.0 µg/mL and diluted in DI water (1:1, v/v)
SPE well plate Discovery DSC-CN 96-well SPE, 100 mg/well (575636-U)
condition 1 mL methanol; 1 mL DI water.
sample addition 1 mL spiked urine
washing 1 mL 20% methanol
elution 1 mL methanol
eluate post-treatment Evaporate eluate with nitrogen at 30 C. Reconstitute in 200 µl mobile phase.
column Discovery HS F5, 5 cm x 4.6 mm I.D., 3 µm particles (567504-U)
column temp. 35 C
mobile phase [A] methanol; [B] DI water; (40:60, A:B)
flow rate 1.5 ml/min
injection 5 µl
detector UV, 240 nm
Description
Categories Analytical Chromatography, Clinical, SPE Applications, Steroids, steroids, Pharmaceutical & Clinical applications
Featured Industry Clinical
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for SPE, application for HPLC