HPLC Analysis of Cosmetic Preservatives on Ascentis® Express C18 (1.2 mL/min)
HPLC Analysis of Cosmetic Preservatives on Ascentis® Express C18 (1.2 mL/min)
Conditions
column Ascentis Express C18, 7.5 cm x 3 mm I.D., 2.7 µm particles (53812-U)
column temp. 35 °C
mobile phase [A] water; [B] methanol; (55:45, A:B)
flow rate 1.2 mL/min
sample methyl-, benzylparaben, 50 mg/L; ethylparaben, 30 mg/L; propyl-, butylparaben, 100 mg/L; 2-phenoxyethanol, 1.5 g/L; 1-phenoxy-2-propanol, 3 g/L; all in 40% methanol
injection 1 µL
detector UV, 250 nm
Description
Categories Analytical Chromatography, Flavors, Fragrances
Featured Industry Cosmetics, Personal Care, and Cleaning Products