Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Coumafuryl Enantiomers on Astec® CHIROBIOTIC® V
HPLC Analysis of Coumafuryl Enantiomers on Astec® CHIROBIOTIC® V
Conditions
column CHIROBIOTIC V, 25 cm x 4.6 mm I.D., 5 µm particles (11024AST)
column temp. 25 C
mobile phase [A] 20 mM ammonium nitrate, pH 5.5; [B] THF; (90:10, A:B)
flow rate 1.2 ml/min
sample 2 mg/mL in methanol
injection 1 µl
detector UV, 254 nm
Description
Categories Analytical Chromatography, Chiral Applications, Pesticides & Herbicides, Environmental, Pesticides, All Environmental Applications
Featured Industry Chemicals and Industrial Polymers
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC