HPLC Analysis of Cresols and Phenol on Astec® CYCLOBOND® I 2000
HPLC Analysis of Cresols and Phenol on Astec® CYCLOBOND® I 2000
Conditions
column CYCLOBOND I 2000, 25 cm x 4.6 mm I.D., 5 µm particles (20024AST)
column temp. 22 °C
mobile phase [A] water; [B] methanol; (90:10, A:B)
flow rate 1 mL/min
sample 0.5 mg/mL in methanolpeak 1: o-cresol (10.70 min)peak 2: phenol (11.70 min)peak 3: m-cresol (14.20 min)peak 4: p-cresol (20.10 min)
injection 1 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Chiral Applications, Phenols, Chemicals and Industrial Polymers, Alcohols, Phenols & Diols
Featured Industry Chemicals and Industrial Polymers
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CYCLOBOND is a registered trademark of Sigma-Aldrich Co. LLC