Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Crufomate (Ruelene) Enantiomers on Astec® CYCLOBOND® I 2000
HPLC Analysis of Crufomate (Ruelene) Enantiomers on Astec® CYCLOBOND® I 2000
Conditions
column CYCLOBOND I 2000, 25 cm x 4.6 mm I.D., 5 µm particles (20024AST)
column temp. ambient
mobile phase [A] acetonitrile; [B] acetic acid; [C] triethylamine; (100:0.2:0.1, A:B:C)
flow rate 1 ml/min
sample 1 mg/mL in methanol
injection 5 µl
detector UV, 254 nm
Description
Categories Analytical Chromatography, Chiral Applications, Environmental, Pesticides & Herbicides, Pesticides, All Environmental Applications
Featured Industry Chemicals and Industrial Polymers
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CYCLOBOND is a registered trademark of Sigma-Aldrich Co. LLC