HPLC Analysis of Cyclophosphamide Enantiomers on Astec® CHIROBIOTIC® T
HPLC Analysis of Cyclophosphamide Enantiomers on Astec® CHIROBIOTIC® T
Conditions
column CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 µm particles (12024AST)
column temp. 23 °C
mobile phase [A] water; [B] ethanol; [C] THF; (90:5:5, A:B:C)
flow rate 0.75 mL/min
sample 1 mg/mL in methanol
injection 5 µL
detector UV, 205 nm
Description
Categories Analytical Chromatography, Chiral Applications, Antineoplastics, Pharmaceutical (small molecule), All Pharmaceutical Applications
Featured Industry Pharmaceutical (small molecule)
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC