HPLC Analysis of Cylert (Pemoline) and Ritalin (Methylphenidate) on Discovery® C18
HPLC Analysis of Cylert (Pemoline) and Ritalin (Methylphenidate) on Discovery® C18
Conditions
column Discovery C18, 15 cm x 4.6 mm I.D., 5 µm particles (504955)
column temp. 35 °C
mobile phase 80:20 water 10 mM ammonium acetate, pH 3.0 with formic acid):acetonitrile
flow rate 1.0 mL/min
sample Cylert (50 µg/mL) and Ritalin (100 µg/mL) in 85:15 water:CH3OH
injection 10 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, ADHD Drugs, AD/HD Drugs
Featured Industry Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC