HPLC Analysis of CYP3A Inhibitors on Ascentis® Express RP-Amide
HPLC Analysis of CYP3A Inhibitors on Ascentis® Express RP-Amide
Conditions
column Ascentis Express RP-Amide, 10 cm x 2.1 mm I.D., 2.7 µm particles (53913-U)
column temp. 35 °C
mobile phase [A] 20 mM ammonium formate in 50:50 (v/v) water:acetonitrile
flow rate 0.6 mL/min
injection 1 µL
detector ESI(+), m/z 100-600
Description
Analysis Note This application demonstrates the suitability of the Ascentis Express RP Amide for the analysis of CYP3A Inhibitors.
Categories Analytical Chromatography, Clinical, Clinical Enzymes, proteins, HPLC Applications Enzymes, Proteins
Featured Industry Clinical