HPLC Analysis of DACH-ACR Enantiomers on Astec® P-CAP™
HPLC Analysis of DACH-ACR Enantiomers on Astec® P-CAP™
Conditions
column Astec (R,R) P-CAP or (S,S) P-CAP, 25 cm x 4.6 mm I.D., 5 µm particles (31024AST, 33024AST)
column temp. 25 °C
mobile phase [A] acetonitrile; [B] methanol; (97:3, A:B)
flow rate 1 mL/min
sample (R,R)- and (S,S)-DACH-ACR (N-(2-acryloylamino-(1R,2R)-cyclohexyl)-acrylamide)
injection 10 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Chiral Applications, Organic Chemicals, Chemicals and Industrial Polymers, Other Organic Compound Applications
Featured Industry Chemicals and Industrial Polymers
Legal Information P-CAP is a trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC