Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Dansyl-Phenylalanine Enantiomers on Astec® CHIROBIOTIC® R
HPLC Analysis of Dansyl-Phenylalanine Enantiomers on Astec® CHIROBIOTIC® R
Conditions
column CHIROBIOTIC R, 25 cm x 4.6 mm I.D., 5 µm particles (13024AST)
column temp. 25 C
mobile phase [A] 20 mM ammonium acetate; [B] methanol; (70:30, A:B)
flow rate 1 ml/min
sample 1 mg/mL in methanol
injection 1 µl
detector UV, 260 nm
Description
Categories Analytical Chromatography, Chiral Applications, Amino Acids, Life Science & Biopharma, All Biochemical Applications, All Amino Acids, Dansylated Amino Acids
Featured Industry Life Science and Biopharma
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC