HPLC Analysis of Dehydroascorbic Acid and Ascorbic Acid (Vitamin C) on SeQuant ZIC-HILIC
HPLC Analysis of Dehydroascorbic Acid and Ascorbic Acid (Vitamin C) on SeQuant ZIC-HILIC
Conditions
column SeQuant ZIC-HILIC, PEEK, 150 x 4.6 mm, 5 µm, 200 Ã… (1.50455.0001)
mobile phase [A] acetonitrile; [B] ammonium acetate; (70:30, A,B)
flow rate 0.5 mL/min
pressure 375 psi (2.6 MPa)
injection 5 µL
detector 240 nm
Description
Featured Industry Vitamins, Nutraceuticals, and Natural Products