Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Derivatized Diisocyanates on Ascentis® Express RP-Amide
HPLC Analysis of Derivatized Diisocyanates on Ascentis® Express RP-Amide
Conditions
column Ascentis Express RP-AMIDE 15 cm x 4.6 mm, 2.7 µm particles (53931-U)
column temp. 26 C
mobile phase [A] 50mM Ammonium Acetate; [B] Acetonitrile
gradient 30% B held for 1.5 min; 30 to 60% B in 10 min; held at 60% B for 5 min
flow rate 0.8 ml/min for 1.5 min; 0.8 ml/min to 1.5 ml/min in 10 min.
sample 125 µg/mL 1-2PP + 10 µg/mL 2,6-TDIP + 10 µg/mL 1,6-HDIP + 10 µg/mL 2,4-TDIP + 10 µg/mL 4,4-MDIP in 70:30 Water:ACN
injection 5 µl
detector UV, 254 nm
Description
Analysis Note This application shows the separation of four common derivatized diisocyanate compounds related to the use of ORBO80 filters on fused core technology.
Categories Analytical Chromatography, Air Monitoring Applications, Isocyanates
Featured Industry Environmental