HPLC Analysis of Diclofenac and Salicylic Acid on Ascentis® C18
HPLC Analysis of Diclofenac and Salicylic Acid on Ascentis® C18
Conditions
column Ascentis C18, 15 cm x 4.6 mm I.D., 5 µm particles (581324-U)
column temp. 35 °C
mobile phase [A] 20 mM phosphoric acid (pH 2.0 unadjusted); [B] acetonitrile; (50:50, A:B)
flow rate 1 mL/min
sample 50 µg/mL each in mobile phase
injection 10 µL
detector UV, 220 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Anti-inflammatories, Organic Acids