HPLC Analysis of Diet Soda Additives on Ascentis® Express C18
HPLC Analysis of Diet Soda Additives on Ascentis® Express C18
Conditions
column Ascentis Express C18, 3 cm x 4.6 mm I.D., 2.7 µm particles (53818-U)
column temp. 40 °C
mobile phase 20% A constant; 75 to 20% B, 5 to 60% C in 1 min; held at 20% B, 60% C for 0.1 min
flow rate 3 mL/min
pressure 3191 psi (220 bar)
sample (Diet Soda 100 - 500 µg/mL in buffer)
injection 1 µL
detector UV, 214 nm
Description
Analysis Note This application demonstrates the suitability of Ascentis Express C18 for the efficient separation of common drink additives used in diet cola.
Categories Analytical Chromatography, Food & Beverage Analysis, Flavors, Fragrances, Beverage Testing, Soft Drinks
Featured Industry Food and Beverages